COMH93D5BAE2 Combi-Blocks, Inc.

3,3'-Dimethoxyazoxybenzene

  • MDL number MFCD01861416
  • Cas Number : 13556-81-5
  • Molecular Weight : 258.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1632

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1632
Availability details for
Quantity
  • Availability
  • Add to Cart