COMH93D5BC23 Combi-Blocks, Inc.

Piperazine, 1-[(2-chloro-5-nitrophenyl)methyl]-4-ethyl-

  • MDL number MFCD01466591
  • Cas Number : 414881-87-1
  • Molecular Weight : 283.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2106

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2106
Availability details for
Quantity
  • Availability
  • Add to Cart