COMH93D5BC2C Combi-Blocks, Inc.

2,4,6-Trimethoxy-3-methylbenzaldehyde

  • MDL number MFCD00017319
  • Cas Number : 81574-58-5
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2115
Availability details for
Quantity
  • Availability
  • Add to Cart