COMH93D5BC62 Combi-Blocks, Inc.

Methyl (3beta,19alpha,20alpha)-16,17-didehydro-10,11-dimethoxy-19-methyloxayohimban-16-carboxylate

  • MDL number MFCD01710270
  • Cas Number : 131-02-2
  • Molecular Weight : 412.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2190

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2190
Availability details for
Quantity
  • Availability
  • Add to Cart