COMH93D5BC6E Combi-Blocks, Inc.

3-Hydroxy lidocaine

  • MDL number MFCD00869530
  • Cas Number : 34604-55-2
  • Molecular Weight : 250.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2207

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2207
Availability details for
Quantity
  • Availability
  • Add to Cart