COMH93D5BC92 Combi-Blocks, Inc.

1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran

  • MDL number MFCD00217003
  • Molecular Weight : 258.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2260

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2260
Availability details for
Quantity
  • Availability
  • Add to Cart