COMH93D5BCD4 Combi-Blocks, Inc.

Lithium metasilicate

  • MDL number MFCD00016179
  • Cas Number : 10102-24-6
  • Molecular Weight : 89.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2364

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2364
Availability details for
Quantity
  • Availability
  • Add to Cart