COMH93D5BD07 Combi-Blocks, Inc.

5-(tert-Butyl)-3-(morpholinomethyl)benzene-1,2-diol

  • MDL number MFCD08856950
  • Cas Number : 25825-31-4
  • Molecular Weight : 265.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2428

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2428
Availability details for
Quantity
  • Availability
  • Add to Cart