COMH93D5BD6C Combi-Blocks, Inc.

Meosuc-ala-ala-pro-val-oh

  • MDL number MFCD00133608
  • Cas Number : 107441-48-5
  • Molecular Weight : 470.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2569

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2569
Availability details for
Quantity
  • Availability
  • Add to Cart