COMH93D5BE19 Combi-Blocks, Inc.

(3-Hydroxyphenyl)acetone

  • MDL number MFCD00210438
  • Cas Number : 64479-84-1
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2873

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2873
Availability details for
Quantity
  • Availability
  • Add to Cart