COMH93D5BE1A Combi-Blocks, Inc.

4,5,6,7-Tetraiodoisobenzofuran-1,3-dione

  • MDL number MFCD00083098
  • Cas Number : 632-80-4
  • Molecular Weight : 651.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2875

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2875
Availability details for
Quantity
  • Availability
  • Add to Cart