COMH93D5BE6E Combi-Blocks, Inc.

1-(6-(Hydroxymethyl)pyridin-2-yl)ethanone

  • MDL number MFCD09029472
  • Cas Number : 135450-43-0
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2998

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2998
Availability details for
Quantity
  • Availability
  • Add to Cart