COMH93D5BEA1 Combi-Blocks, Inc.

N-tert-Butyl-2,2,2-trifluoroacetamide

  • MDL number MFCD00626222
  • Cas Number : 1960-29-8
  • Molecular Weight : 169.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3081

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3081
Availability details for
Quantity
  • Availability
  • Add to Cart