COMH93D5BF01 Combi-Blocks, Inc.

1-(4-Phenylpiperazin-1-yl)ethanone

  • MDL number MFCD00100137
  • Cas Number : 21557-13-1
  • Molecular Weight : 204.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3222

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3222
Availability details for
Quantity
  • Availability
  • Add to Cart