COMH93D5BF84 Combi-Blocks, Inc.

2-Methoxy-4,6-dimethylbenzoic acid

  • MDL number MFCD01194293
  • Cas Number : 96881-90-2
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3413

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3413
Availability details for
Quantity
  • Availability
  • Add to Cart