COMH93D5BFD9 Combi-Blocks, Inc.

5-(Pyridin-4-yl)-1h-1,2,4-triazol-3-amine

  • MDL number MFCD01678746
  • Molecular Weight : 161.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3551

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3551
Availability details for
Quantity
  • Availability
  • Add to Cart