COMH93D5C04A Combi-Blocks, Inc.

4-[(3-Methylpyridin-2-yl)oxy]aniline

  • MDL number MFCD06825433
  • Cas Number : 901924-13-8
  • Molecular Weight : 200.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3727
Availability details for
Quantity
  • Availability
  • Add to Cart