COMH93D5C05F Combi-Blocks, Inc.

4-(1H-Benzimidazol-2-yl)-1-(4-fluorophenyl)pyrrolidin-2-one

  • MDL number MFCD06751952
  • Cas Number : 876712-94-6
  • Molecular Weight : 295.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3761

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3761
Availability details for
Quantity
  • Availability
  • Add to Cart