COMH93D5C06C Combi-Blocks, Inc.

2,5-Dioxopyrrolidin-1-yl 6-(2-(propan-2-ylidene)hydrazinyl)nicotinate

  • MDL number MFCD06411383
  • Cas Number : 362522-50-7
  • Molecular Weight : 290.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3779

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3779
Availability details for
Quantity
  • Availability
  • Add to Cart