COMH93D5C098 Combi-Blocks, Inc.

2-Amino-n-(2,4-dichlorophenyl)benzamide

  • MDL number MFCD00175993
  • Cas Number : 34490-00-1
  • Molecular Weight : 281.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3846

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3846
Availability details for
Quantity
  • Availability
  • Add to Cart