COMH93D5C0AD Combi-Blocks, Inc.

N,N'-Bis-(4-chlorobenzylidene)-c-phenylmethanediamine

  • MDL number MFCD01861481
  • Cas Number : 320734-42-7
  • Molecular Weight : 367.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3878

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3878
Availability details for
Quantity
  • Availability
  • Add to Cart