COMH93D5C0B5 Combi-Blocks, Inc.

1-(4-Hydroxy-3-nitrophenyl)propan-2-one

  • MDL number MFCD03410478
  • Cas Number : 1314934-53-6
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3891

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3891
Availability details for
Quantity
  • Availability
  • Add to Cart