COMH93D5C0CD Combi-Blocks, Inc.

Butamirate citrate

  • MDL number MFCD00315222
  • Cas Number : 18109-81-4
  • Molecular Weight : 499.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3929

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3929
Availability details for
Quantity
  • Availability
  • Add to Cart