COMH93D5C111 Combi-Blocks, Inc.

Daunomycinone

  • MDL number MFCD01713208
  • Cas Number : 21794-55-8
  • Molecular Weight : 398.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4052

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4052
Availability details for
Quantity
  • Availability
  • Add to Cart