COMH93D5C11E Combi-Blocks, Inc.

Cytidine 3'-monophosphate

  • MDL number MFCD00006543
  • Cas Number : 84-52-6
  • Molecular Weight : 323.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4073
Availability details for
Quantity
  • Availability
  • Add to Cart