COMH93D5C120 Combi-Blocks, Inc.

6-Methoxy-2-methyl-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD01717130
  • Cas Number : 54893-54-8
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4075
Availability details for
Quantity
  • Availability
  • Add to Cart