COMH93D5C131 Combi-Blocks, Inc.

3-Deoxy-d-galactose

  • MDL number MFCD08457745
  • Cas Number : 4005-35-0
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4098

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4098
Availability details for
Quantity
  • Availability
  • Add to Cart