COMH93D5C1DA Combi-Blocks, Inc.

Isoguanosine hydrate

  • MDL number MFCD01863406
  • Cas Number : 359436-55-8
  • Molecular Weight : 301.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4351

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4351
Availability details for
Quantity
  • Availability
  • Add to Cart