COMH93D5C1DD Combi-Blocks, Inc.

Z-Leu-tyr-chloromethylketone

  • MDL number MFCD00238476
  • Cas Number : 56979-35-2
  • Molecular Weight : 460.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4354

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4354
Availability details for
Quantity
  • Availability
  • Add to Cart