COMH93D5C1EB Combi-Blocks, Inc.

3,4,5-Tribromotoluene

  • MDL number MFCD00075798
  • Cas Number : 73557-59-2
  • Molecular Weight : 328.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4382

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4382
Availability details for
Quantity
  • Availability
  • Add to Cart