COMH93D5C204 Combi-Blocks, Inc.

Amodiaquine

  • MDL number MFCD00552927
  • Cas Number : 86-42-0
  • Molecular Weight : 355.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4422

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4422
Availability details for
Quantity
  • Availability
  • Add to Cart