COMH93D5C230 Combi-Blocks, Inc.

Niclofolan

  • MDL number MFCD00867407
  • Cas Number : 10331-57-4
  • Molecular Weight : 345.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4485

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4485
Availability details for
Quantity
  • Availability
  • Add to Cart