COMH93D5C25C Combi-Blocks, Inc.

S-2-Benzothiazolyl 2-amino-alpha-(methoxyimino)-4-thiazolethiolacetate

  • MDL number MFCD00129148
  • Cas Number : 94088-75-2
  • Molecular Weight : 350.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4547

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4547
Availability details for
Quantity
  • Availability
  • Add to Cart