COMH93D5C264 Combi-Blocks, Inc.

Alpha-l-idopyranose, pentaacetate

  • MDL number MFCD03453193
  • Cas Number : 2152-77-4
  • Molecular Weight : 390.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4558

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4558
Availability details for
Quantity
  • Availability
  • Add to Cart