COMH93D5C3B0 Combi-Blocks, Inc.

Kaempferol 3-o-sophoroside

  • MDL number MFCD01662713
  • Cas Number : 19895-95-5
  • Molecular Weight : 610.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5045
Availability details for
Quantity
  • Availability
  • Add to Cart