COMH93D5C48A Combi-Blocks, Inc.

Boc-val-leu-lys-amc acetate salt

  • MDL number MFCD03788111
  • Cas Number : 73554-84-4
  • Molecular Weight : 675.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5375

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5375
Availability details for
Quantity
  • Availability
  • Add to Cart