COMH93D5C4B7 Combi-Blocks, Inc.

5-Heptyn-3-ol

  • MDL number MFCD00046680
  • Cas Number : 19781-82-9
  • Molecular Weight : 112.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5437

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5437
Availability details for
Quantity
  • Availability
  • Add to Cart