COMH93D5C508 Combi-Blocks, Inc.

Ethyl 2-(3,4-dihydroxyphenyl)acetate

  • MDL number MFCD00564055
  • Cas Number : 83112-21-4
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5554
Availability details for
Quantity
  • Availability
  • Add to Cart