COMH93D5C524 Combi-Blocks, Inc.

Z-Gly-leu-nh2

  • MDL number MFCD00070668
  • Cas Number : 7535-72-0
  • Molecular Weight : 321.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5596
Availability details for
Quantity
  • Availability
  • Add to Cart