COMH93D5C531 Combi-Blocks, Inc.

5-O-Benzoyl-1,2-di-o-acetyl-3-deoxy-d-ribofuranose

  • MDL number MFCD01630915
  • Cas Number : 4613-71-2
  • Molecular Weight : 322.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5614

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5614
Availability details for
Quantity
  • Availability
  • Add to Cart