COMH93D5C573 Combi-Blocks, Inc.

3-(2,4-Difluorophenyl)prop-2-yn-1-ol

  • MDL number MFCD06803802
  • Cas Number : 288100-99-2
  • Molecular Weight : 168.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5725

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5725
Availability details for
Quantity
  • Availability
  • Add to Cart