COMH93D5C586 Combi-Blocks, Inc.

Ethyl 4,6-difluoro-1h-indole-2-carboxylate

  • MDL number MFCD07781308
  • Cas Number : 870536-93-9
  • Molecular Weight : 225.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5754
Availability details for
Quantity
  • Availability
  • Add to Cart