COMH93D5C592 Combi-Blocks, Inc.

4-Methoxy-6-methyl-2h,5h,6h,7h,8h-[1,3]dioxolo[4,5-g]isoquinoline

  • MDL number MFCD00868501
  • Cas Number : 550-10-7
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5769

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5769
Availability details for
Quantity
  • Availability
  • Add to Cart