COMH93D5C619 Combi-Blocks, Inc.

N-Cyclopentylpyrazin-2-amine

  • MDL number MFCD01471625
  • Cas Number : 420844-66-2
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5965

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5965
Availability details for
Quantity
  • Availability
  • Add to Cart