COMH93D5C61A Combi-Blocks, Inc.

5-(3-Fluorophenyl)nicotinaldehyde

  • MDL number MFCD06410218
  • Cas Number : 887973-72-0
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5966
Availability details for
Quantity
  • Availability
  • Add to Cart