COMH93D5C629 Combi-Blocks, Inc.

2,2-Dihydroxymalonic acid

  • MDL number MFCD00085305
  • Cas Number : 560-27-0
  • Molecular Weight : 136.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5994

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5994
Availability details for
Quantity
  • Availability
  • Add to Cart