COMH93D5C638 Combi-Blocks, Inc.

Nonyl gallate

  • MDL number MFCD00016433
  • Cas Number : 10361-11-2
  • Molecular Weight : 296.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6015

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6015
Availability details for
Quantity
  • Availability
  • Add to Cart