COMH93D5C669 Combi-Blocks, Inc.

4,4'-Diacetoxystilbene

  • MDL number MFCD00017233
  • Cas Number : 63449-52-5
  • Molecular Weight : 296.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6094

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6094
Availability details for
Quantity
  • Availability
  • Add to Cart