COMH93D5C691 Combi-Blocks, Inc.

Tetrakis(hydroxymethyl)phosphonium sulfate

  • MDL number MFCD00054243
  • Cas Number : 55566-30-8
  • Molecular Weight : 252.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6163

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6163
Availability details for
Quantity
  • Availability
  • Add to Cart