COMH93D5C6B9 Combi-Blocks, Inc.

Dl-alpha-methyl-m-tyrosine

  • MDL number MFCD00021681
  • Cas Number : 305-96-4
  • Molecular Weight : 195.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6221

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-6221
Availability details for
Quantity
  • Availability
  • Add to Cart